VWR® BaseBond Micro Vials with Crimp Neck, ND11
Catalog # 548-2648A
Supplier: VWR Collection
Specifications
- Ø×A:11,6×32,0 mm
- Confezione:100 per pack
- Capacità:0,20 ml
- Colore:ambra
- Descrizione:Crimp neck vial with integrated microinsert
- Materiale:Vetro
- Conf:1.000
Specifications
About this item
- 1st hydrol. class, writing patch, with small micro-insert
- BaseBond