VWR® Short Thread Vials ND9
Short thread vial with integrated micro-insert, 0,20 ml, 11,6×32,0 mm, Vetro, trasparente, 100 per pack
Codice catalogo 548-2644A
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
The vials can be used on all common autosamplers due to their technical geometry.
- 1st hydrol. class, label and filling lines
- Top bonded
Specifiche
- Ø×A:11,6×32,0 mm
- Confezione:100 per pack
- Capacità:0,20 ml
- Colore:trasparente
- Descrizione:Short thread vial with integrated micro-insert
- Materiale:Vetro