ReproSil 80 C6 HPLC Columns
Supplier: DR MAISCH
Certificates
About this item
ReproSil 80 are type A silica which can replace Spherisorb columns. It has been developed to provide the same behavior of waters Sherisorb columns. It represents a competitive alternative with a very high reproducibility.
Specifications
- Catalog No:
- MAISR05.60.S1546
- MAISR03.60.S1246
- MAISR05.60.S1246
- Carico di carbonio:
- 6%
- 6%
- 6%
- Con endcapping:
- Sì
- Sì
- Sì
- Range di pH:
- 2 - 8
- 2 - 8
- 2 - 8
- Area superficiale:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- Codice USP:
- L15
- L15
- L15