Filter Fleece
Filter fleece
Codice catalogo INFIPPV75-37-100
Nome del fornitore: PIEPER FILTER
Vai a
Product Details & Documents
Filter fleece is suitable for drainage under membranes. It is also used in chipping processes to keep the coolant free of the chips. The fleece is also suitable for coarse air filtration.
- Polypropylene fleece
- Thickness: 75 µm
- Sampling according to IFA 7512, IFA 6172
Specifiche
- Descrizione:Filter fleece