The TNP (Mouse) IgG ELISA kit
Catalog # ABNOKA6743.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Conf:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:TNP
Specifications
About this item
The TNP (Mouse) IgG ELISA Kit is based on a solid phase enzymelinked immunosorbent assay (ELISA). The assay uses TNP-BSA as the capture antigen (coated on the microtiter wells) and (HRP) conjugated anti-mouse IgG antibodies for detection.