Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgE Mouse Recombinant Antibody [clone: TES-C21]
Anti-IgE Mouse Recombinant Antibody [clone: TES-C21]
Catalog # ABNORAB01167.200UG
Supplier:  Abnova
Anti-IgE Mouse Recombinant Antibody [clone: TES-C21]
Catalog # ABNORAB01167.200UG
Supplier:  Abnova

Specifications

  • Simbolo dell'antigene:
    IgE
  • Clonalità:
    Recombinant
  • Clone:
    TES-C21
  • Ospite:
    Mouse
  • Reattività:
    Human
  • Immunogeno:
    Recombinant mouse monoclonal antibody raised against human IgE.
  • Conf:
    200 µG

Specifications

About this item