Protein quantitation assay kits, Non-Interfering™ (NI™) Protein Assay
Codice catalogo 786-005
Nome del fornitore: G-Biosciences
Numero CAS:
Specifiche
- Conf:1 KIT
- Descrizione:Non-Interfering™ (NI™) Protein Assay with albumin standard, 500 assays
Specifiche
Informazioni su questo articolo
A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal protein-to-protein variation. The assay is unaffected by the presence of common laboratory agents such as reducing agents, chelating agents, detergents, amines, sugars, chaotropes, salts, drugs, antibiotics, cobalt, and others.