Hooded Softshell Jackets, KX362 - KX3
: Portwest
The KX3 hooded softshell jackets is precision engineered using a revolutionary breathable, waterproof and windproof ripstop fabric.
- Ripstop fabric to ensure durability and resistance to tearing and ripping
- Contemporary design with an ergonomic cut
- Side zip pockets, Hook and loop cuffs for a secure fit, grown on hood is stylish and practical
- Reflective piping for increased visibility, drawcord adjustable hem, curved back hem for added protection
- Easy grip pullers attached to all zips
- Chin guard for added comfort and stability
- Four pockets for ample storage, has phone pocket
Its sports inspired design includes a longer back length, dynamic shaped sleeves, adjustable cuffs, reflective trims and multiple secure pockets for ample storage. An ideal choice for the modern worker.
Ergonomically shaped hood for a perfect fit. Water resistant eco-conscious fabric finish, water beads away from fabric surface.
Shell fabric: Ripshell: 92% polyester, 8% elastane ripstop laminated to 100% polyester waffle micro fleece (3 L) 310 g
Standards: Waterproof/Breathability WP 8000 mm, MVP 3000 g/M²/24 h
: Do not bleach, dry clean, or tumble dry.
- Catalog No:
- PRTWKX362BGYS
- PRTWKX362BGYM
- PRTWKX362BGYXL
- PRTWKX362BGYXXL
- PRTWKX362BGYXXXL
- PRTWKX362BGYL
- Modello:
- KX362 - KX3
- KX362 - KX3
- KX362 - KX3
- KX362 - KX3
- KX362 - KX3
- KX362 - KX3
- Applicazioni per abiti:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 92% polyester, 8% elastane
- 92% polyester, 8% elastane
- 92% polyester, 8% elastane
- 92% polyester, 8% elastane
- 92% polyester, 8% elastane
- 92% polyester, 8% elastane
Frequently Bought Together