Configurator for Gas Detection System, X-am® 2800
Codice catalogo DRAE3703900
Nome del fornitore: Dräger
Specifiche
- Descrizione:Drager X-am 2800 configurator
- Conf:1
Specifiche
Informazioni su questo articolo
Configurator for Dräger X-am® 2800 consisting of basic device with data logger, delivered with manufacturer and calibration certificate.
- Configurator: up to 4 sensors can be selected, sensor configuration can be changed later in product life
Customer-specific settings (firmware) can be selected when ordering, examples: Measurement gas for the Ex sensor (factory setting: methane), alarm thresholds, alarm behaviour, quick menu options, D-light etc.
Ordering information: Power supply (battery) optional (factory setting: with battery), without charger.