VERISHIELD™ Earmuffs 100 IS
Catalog # DALL1035101-VS
Supplier: HONEYWELL SAFETY
Specifications
- Altezza ripiano:25
- Descrizione:VS140 VeriSheild™ earmuff without air flow control, low level/flat, over the head
- Conf:10
Specifications
About this item
VeriSheild™ is ideal for wearing with other PPE which may be required in particular environments. They offer a choice of styles and attenuation (SNR) levels makes it easy to find the suitable earmuffs for an appropriate level of protection, and comfort.
- Easy to fit
- Suitable for wearers with sensitive ear canals
- Suitable for wearing with hearing aids
- It helps to keep ears warm
Air flow control and sound management technology that improves employee safety by blocking harmful noise while allowing voice and signals heard more naturally.