Tube holder S
Catalog # BINH1014525
Supplier: 3B Scientific
Specifications
- Descrizione:Tube Holder S
- Conf:1
Specifications
About this item
Tube holder to support all S series electron tubes for easy and safe operation. The five pin sockets for the tube are concealed inside the tube holder. A cathode protection switch is integrated into the tube holder, to protect the heated cathode from excessive voltage. The base plate has a slot for attaching the Helmholtz pair of coils S.
- Terminals : 4 mm safety sockets
- Dimensions: approx. 130×190×250 mm³
- Weight: approx. 570 g