Ready-to-Load™ kits, the mystery of the crooked cell
Catalog # 763-0010
Supplier: EDVOTEK
Specifications
- Descrizione:The Mystery of the Crooked Cell
- Conf:1
Specifications
About this item
This simple experiment demonstrates detection of the mutation that causes Sickle Cell Anaemia. In this simulation, students use electrophoresis to separate dyes that represent patient samples and controls.
- For 10 lab groups
- Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.