Ready-to-Load™ kits, why do people look different?
Catalog # 763-0007
Supplier: EDVOTEK
Specifications
- Descrizione:Why Do People Look Different?
- Conf:1 KIT
Specifications
About this item
This kit is for teaching students how people's physical traits are a reflection of their genes. In this simulation, students use electrophoresis to separate dyes which represent genetic traits.
- For 10 lab groups
- Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.