How Do You Clone a Gene?
How Do You Clone a Gene?
Codice catalogo EDVOS-20
Nome del fornitore: EDVOTEK
Vai a
Product Details & Documents
In this kit, a set of multi-coloured links demonstate a variety of molecular biology simulations. Students learn about digesting DNA with restriction enzymes, cloning genes in plasmids, protein structure, and more.
- For 10 lab groups
- Complete in 30 minutes
Informazioni per l'ordine: Kit includes instructions, molecular biology models, small plastic bags.
Specifiche
- Descrizione:How Do You Clone a Gene?