What Does DNA Look Like?
What Does DNA Look Like?
Codice catalogo EDVOS-10
Nome del fornitore: EDVOTEK
Vai a
Product Details & Documents
This lab activity shows students what real chromosomal DNA looks like and allows them to explore the procedures involved in DNA extraction.
- For 10 lab groups
- Complete in 30 minutes
Also required: 95% ethanol or isopropyl alcohol.
: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, minilinks, glass rod, DNA spooling rods, test tubes, and salt.
Specifiche
- Descrizione:What Does DNA Look Like?