Ready-to-Load™ kits, in search of the cancer gene
N° de catalogue EDVO314
Fournisseur: EDVOTEK
Spécifications
- Descrizione:In Search of the Cancer Gene
- Conf:1
Spécifications
À propos de cet article
Suppressor genes, such as p53, are essential for cell functions. Mutations in the p53 gene can be correlated to predisposition for certain cancers. Mutations of genes can either be inherited or accumulated due to environmental insults. This experiment deals with a family pedigree determination of several generations relating to cancer formation due to p53 gene mutation.
- For 6 lab groups
- Complete in 60 minutes
Also required: Electrophoresis tank, power supply, automatic micropipette with tips, 65 °C water bath, and UV transilluminator.
Livraison: Kit includes instructions, Ready-to-Load™ predigested DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® Ethidium Bromide, pipette, and 5 autoradiograms.