Transformation with green fluorescent protein
Codice catalogo EDVO223/AP08
Nome del fornitore: EDVOTEK
Specifiche
- Descrizione:Transformation with Green Fluorescent Protein
- Conf:1
Specifiche
Informazioni su questo articolo
In this experiment, transformed cells take up a plasmid containing the Green Fluorescent Protein (GFP) gene. The GFP gene was isolated from the jellyfish Aequorea victoria. Transformed colonies expressing the GFP protein are visibly green in normal light but will fluoresce brightly when exposed to longwave UV light.
- For 10 lab groups
- Plating 5 minutes; incubation overnight; transformation efficiency 15 minutes
Also required: 37 °C incubator, water bath, longwave UV lamp.
Delivery: Kit includes instructions, cells, plasmid DNA, IPTG, ampicillin, transformation solution, ReadyPour agar, Luria broth, petri dishes, sterile pipettes, loops and microtubes.