LMDA (Lee's Multi-Differential Agar)
LMDA (Lee's Multi-Differential Agar), 500 g, Plastic bottle, 1
Nome del fornitore: DOEHLER GROUP
LMDA (Lee's Multi-Differential Agar)
Codice catalogo DOEH923555244
Nome del fornitore: DOEHLER GROUP
Numero CAS:
Vai a
Product Details & Documents
Siebel-Instituts agar medium for the selective detection and quantitative determination of microorganisms (bacteria, yeasts) from the brewing environment.
- Certified by the Siebel-Institute for technology
- Ready-to-use powder for easy handling
- Functionality tested with strains originally isolated from the brewing environment
Identification of the target bacteria is facilated by characteristic colour changes of the medium. In addition, acid-forming bacteria show a typical halo formation due to the breakdown of the calcium carbonate contained in the LMDA medium.
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Tipo di conf.:Plastic bottle
- Conf:1
- Descrizione:LMDA (Lee's Multi-Differential Agar)
- Riempimento:500 g