Stationary Tank System for Lubricants, MULTI-Tank
CEMG7536
: CEMO
- Capacità:400 L
- Descrizione:MULTI-Tank stationary tank system, HDPE
- L×P×A:740×730×1170 mm
- Conf:1
Tank from HDPE with general construction inspection approval with rigidly connected sheet steel pallet for handling with a forklift or pallet truck from any of the four sides.
- Tank as described above with integral sheet steel pallet for driving under with a forklift or hand forklift from any of four sides
- Approved for transport according to the ADR, but without a pump fitted
- Lower centre of gravity increases stability and improves handling/ transport
- For storage of diesel, biodiesel without a bund, even in water-protected areas
- Certified for installation indoors
Individual tank with integral galvanised sheet steel bund, standard accessories such as filling contents gauge, optical leakage detector device, integral carrying handle, no pump.