96 Well Plate, RCH PWCX
Codice catalogo TCAT675-0201RCH
Nome del fornitore: Tecan
Specifiche
- Descrizione:Plate RCH PWCX, 1 ml, 19 µm
- Fase:PWCX
- Riempimento:20 mg
- Conf:1
Specifiche
Informazioni su questo articolo
Polymeric, weak cation exchange chemistry for strong bases.
- Ion-exchange
- Liquid handler compatible
A 19 µm polymer micro particulate with weak cationic exchange chemistry providing large contac zone for sorbent analyte interaction resulting in a fast and uniform sample prep.