Pipette jar
Pipette jar, white PE, Ø 152 × 460 mm (6 × 18")
Codice catalogo BELAF171300018
Nome del fornitore: Bel-Art Products, a Part of SP
Vai a
Product Details & Documents
Store and soak pipettes in these leakproof, molded polyethylene pipette jars.
Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate.
Specifiche
- Descrizione:Pipette jar, white PE, Ø 152 × 460 mm (6 × 18")