Orders LinkContactUsLinkComponent
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic battery crimping tools effortlessly crimp/decap aluminum and 2-part aluminum/steel caps with just the push of a button.
  • Consistently crimp caps to a reproducible seal setting with less physical strain
  • Powerful brushless motor with a fast cycle
  • Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
  • Innovative charging system, which includes a warning light when recharging is necessary
  • Can be operated while charging
  • The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
  • OLED screen for simple operation displays tool size, battery status, crimp setting and cycle result
  • Performance tracking through an internal cycle log
Tools in 8, 11, 13, and 20 mm crimping and decapping.
Product Family Options
Crimpers
  • Descrizione
  • Electronic crimper tool for 8 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic crimper tool for 11 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic crimper tool for 13 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic flip off crimping tool for 13 mm flip off crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic crimper tool for 20 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic flip off crimping tool for 20 mm flip off crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
Decappers
  • Descrizione
  • Electronic decapper tool for 11 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic decapper tool for 13 mm crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • Electronic decapper tool for 20 mm aluminum crimp caps
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
Accessories
  •  
    Descrizione
  • SGEA5ASTA0
    Stand for CRS electronic and hand crimpers and decappers
    1 * 1 ST - SFr. 84.00
  •  
    Descrizione
  • SGEA5ABAS0
    Base for electronic crimping tool and high powered crimping tool
    1 * 1 ST - SFr. 278.00