ContactUsLinkComponent
Whatman™ Reaction Folders, Sealable Hybridisation Bags
Whatman™ Reaction Folders, Sealable Hybridisation Bags
Codice catalogo WHAT10483064
Nome del fornitore:  Whatman products (Cytiva)
undefined
Whatman™ Reaction Folders, Sealable Hybridisation Bags
Codice catalogo WHAT10483064
Nome del fornitore:  Whatman products (Cytiva)
Specifiche
  • Descrizione:
    Reaction folders
  • Conf:
    50
  • L×L:
    254×203 mm
Specifiche
Informazioni su questo articolo
Whatman reaction folders are sealable bags for hybridisation and incubation reactions.
  • Folders can be sealed with a standard heat-sealer
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridisation bags.