First Aid Station
Product Details & Documents
A First Aid station containing a variety of first aid products, intended to be used in first aid situations.
- Adhesive bandages (plasters) and beige soft foam bandage are placed in dispensers for easy access and theft-proof storage
- Products are easily available and protected against dust and moisture by transparent lids
- Space for additional products is provided behind a lid with first aid instructions
- A special key is provided for refill replacement
Medical devices in the station are CE-labelled. The station contains >70% post-consumer recycled plastic (>85% in green coloured parts).
Contents:
2× Cederroth 4-in-1 Bloodstopper dressings
3× Cederroth 4-in-1 mini Bloodstopper dressings
1× 2 Cederroth burn gel dressings
1× Cederroth eye and wound cleansing spray 150 ml
1× Salvequick plaster dispenser including 45 plastic plasters
40× textile plasters
1× Soft Foam dispenser including 6 cm x 2 m Soft Foam bandage beige
1× 20 Salvequick wound cleansers
1× Cederroth protection kit
1× First aid instructions
1× Refill key
Colour: Green.
W×D×H (mm) 290×120×560.
Specifiche
- Descrizione:First aid station
- Cat. No.:CEDE51011026
- Environmentally Preferable:Materiali sostenibili|Imballi sostenibili
- Certificazione ambientale:Verification of recycled plastic material by 3rd party, see separate document 'Cederroth Stations recycled plastic'