Doypack® Paper/PET/CPP Paper Bags
Informazioni su questo articolo
The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.
- Food contact
- Available in different sizes
- Ideal for wet and dry content
- Doypack® packaging
- Ideal for shelf display
- Available in stock
The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag is reclosable and reusable. All Doypack® Stand Up pouches in stock are equipped with a Clicky® closure system, with tactile & sound effects for an intuitive and sensory closure.
5 varianti
Product Details & Documents
The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.
- Food contact
- Available in different sizes
- Ideal for wet and dry content
- Doypack® packaging
- Ideal for shelf display
- Available in stock
The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag is reclosable and reusable. All Doypack® Stand Up pouches in stock are equipped with a Clicky® closure system, with tactile & sound effects for an intuitive and sensory closure.
Product Family Options
Informazioni sul prodotto
- CapacitàMaterialeSpessoreL×LDisponibilitàPrice
- 129-3901Doypack® Paper/PET/CPP bags, 2×25 mm bottom gussetDoypack® Paper/PET/CPP bags, 2×25 mm bottom gusset100 mlPET/CPPPAPER40/PET12/CPP6085×125 mmLog in per disponibilità1 * 500 STSFr. 312.00
- 129-3902Doypack® Paper/PET/CPP bags, 2×32 mm bottom gussetDoypack® Paper/PET/CPP bags, 2×32 mm bottom gusset250 mlPET/CPPPAPER40/PET12/CPP60110×160 mmLog in per disponibilità1 * 500 STSFr. 349.00
- 129-3903Doypack® Paper/PET/CPP bags, 2×35 mm bottom gussetDoypack® Paper/PET/CPP bags, 2×35 mm bottom gusset500 mlPET/CPPPAPER40/PET12/CPP60130×190 mmLog in per disponibilità1 * 500 STSFr. 384.00
- 129-3904Doypack® Paper/PET/CPP bags, 2×40 mm bottom gussetDoypack® Paper/PET/CPP bags, 2×40 mm bottom gusset750 mlPET/CPPPAPER40/PET12/CPP60160×230 mmLog in per disponibilità1 * 500 STSFr. 436.00
- 129-3905Doypack® Paper/PET/CPP bags, 2×45 mm bottom gussetDoypack® Paper/PET/CPP bags, 2×45 mm bottom gusset1000 mlPET/CPPPAPER40/PET12/CPP60180×255 mmLog in per disponibilità1 * 500 STSFr. 498.00