Sentinel XE® LiPO Rechargeable Battery
Catalog # ILCDS-3503
Supplier: ILC DOVER
Specifications
- Descrizione:Sentinel XE® LiPO rechargeable battery
- Conf:1
Specifications
About this item
12 hour rechargeable lithium polymer battery.
- Quick/secure snap in design
- Lithium polymer technology
The Sentinel XE® LiPO rechargeable battery provides 12 hours of service on a single charge.
Certifications: Approved to EN 62133-2:2017.