Sentinel® Flow Meter
Catalog # ILCDS-2010
Supplier: ILC DOVER
Specifications
- Descrizione:Flow meter
- Seq:04
- Conf:1
Specifications
About this item
Measures the air flow coming out of the blower to help ensure maximum protection.
- Easy to use/read
- Confirms proper flow rate is being achieved