Polyethylene Adhesion Tape
Polyethylene adhesion tape, rosso, 2"
Codice catalogo UTAP1110RE200-P3D
Nome del fornitore: ULTRATAPE
Vai a
Product Details & Documents
UltraTape 1110 is designed specifically for sealing wafer shipping boxes as well as cleanroom packaging and bag sealing. Its specially formulated medium-high adhesive leaves no residue or ghosting on plastic boxes, even after prolonged storage.
Specifiche
- Larghezza:2"
- Colore:rosso
- Descrizione:Polyethylene adhesion tape