ratiolab® Pipette Tips, Colourless
1 - 10 ml, -, Clear
Codice catalogo RATI2400642
Nome del fornitore: RATIOLAB
Vai a
Product Details & Documents
ratiolab® pipette tips are suitable for all commonly used pipette types.
- Precision in each and every detail
- Guarantee for highest quality
- Made of high quality polypropylene
- Autoclavable
- Excellent transparency due to high quality surfaces
Specifiche
- Sterili:-
- Volume:1 - 10 ml
- Colore puntale:Clear