pAAV-AC-GFP AAV Gene Expression Vector
Catalog # ORIGPS100090
Supplier: OriGene
CAS Number:
Specifications
- Conf:10 µG
- Descrizione:pAAV-AC-GFP, AAV vector with C-terminal tGFP tag
Specifications
About this item
AAV Gene Expression Vectors with C-terminal tGFP tag.
- OriGene's TrueORF clones can be shuttled into this vector via PrecisionShuttle, a simple cut-and-ligation process
- The ORF cloned in this vector can be expressed containing c-terminal tGFP
- BGI_F Primer can be used to sequence the target sequence after cloning
- AC_GFP_R Primer can be used to sequence the target sequence after cloning
E. coli Selection: Ampicillin (100 µg/ml)