Order Entry
Switzerland
ContactUsLinkComponent
Anti-Rabbit Monoclonal Antibody [clone: 003] (Biotin)
Anti-Rabbit Monoclonal Antibody [clone: 003] (Biotin)
  NOVUNBP2-90059B
Anti-Rabbit Monoclonal Antibody [clone: 003] (Biotin)
  NOVUNBP2-90059B

 

  • Tipo di anticorpi:
    ULBP-2
  • Clonalità:
    Monoclonale
  • Clone:
    003
  • Coniugazione:
    Biotina
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Reattività:
    Human
  • ID gene:
    ULBP2
  • Sinomini antigene:
    retinoic acid early transcript 1 H|UL16-binding protein 2|NKG2DL2|ULBP2|N2DL2|UL16 binding protein 2|Retinoic acid early transcript 1H|N2DL-2|ALCAN-alpha|RAET1H|RAET1HNKG2DL2|NKG2D ligand 2
  • Purezza:
    Protein A purified
  • Tampone di conservazione:
    PBS
  • Temperatura di conservazione:
    Store at 4 ˚C. Preserve with 0,05% sodium azide.
  • Immunogeno:
    This antibody was obtained from a rabbit immunised with purified, recombinant human ULBP-2 (Uniprot#: Q9BZM5; Met1-Ser217).
  • Conf:
    0,1 mL

 

 

Ligand for the NKG2D receptor, together with at least ULBP1 and ULBP3. ULBPs activate multiple signalingpathways in primary NK cells, resulting in the production of cytokines and chemokines. Binding of ULBPs ligands toNKG2D induces calcium mobilization and activation of the JAK2, STAT5, ERK and PI3K kinase/Akt signal transductionpathway. In CMV infected cells, interacts with soluble CMV glycoprotein UL16. The interaction with UL16 blocked theinteraction with the NKG2D receptor, providing a mechanism by which CMV infected cells might escape the immune system.UL16 also causes ULBP2 to be retained in the ER and cis-Golgi apparatus so that it does not reach the cell surface.

 : Primary antibodies are guaranteed for 1 year from date of receipt.
 : This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type:
Antigen:
Clonality: Monoclonal
Clone: 003
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human