rubis® Tweezers Set
Catalog # OUTI1K531
Supplier: OUTILS RUBIS
Specifications
- Descrizione:5-piece tweezers set
- Materiale:Acciaio inossidabile
- Conf:5
Specifications
About this item
Set of 5 tweezers containing 1 tweezer 00-SA, 1 tweezer 2A-SA, 1 tweezer 3C-SA, 1 tweezer 5-SA, and 1 tweezer 7-SA.
- Made from stainless steel
- Acid-resistant
- Resistant against hydrogen fluoride and nitric acid
- 100% anti-magnetic
- Packaged in a practical hardcover case
Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology, and electron microscopy.