Chemist's Spoon
150 mm
Codice catalogo USBE3406
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Single-ended chemical spoon made from stainless steel.
- Autoclavable
- Easy to clean
- 16 mm width
- 12 g weight
Excellent chemical resistance. High temperature resistance.
Specifiche
- Lunghezza:150 mm