Notrax® 562 Sanitop® Floor Mattings
Codice catalogo JUSU562S0035BL
Nome del fornitore: JUSTRITE MANUFACTURING
Specifiche
- Colore:nero
- Descrizione:Rubber mat
- L×L:1520×910 mm
- Spessore:13 mm (½")
- Conf:1
Specifiche
Informazioni su questo articolo
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
- Durable rubber compound
- Molded alternating large/small hole drainage system keeps moisture and debris free from work area
- Raised pattern surrounds large holes to channel drainage and provide worker traction
- Beveled ramps on all 4 sides
- 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards. Sanitop® offers a simple, economical solution for wet environments in many industrial or food service applications.
Weight: 9,97903 kg