Regenerating hand and face cream, Physioderm® Creme
Catalog # FAWE14009003
Supplier: FAWECO HAUTSCHUTZ
Specifications
- Descrizione:Physioderm® Creme, 100 ml tube
- Conf:12
Specifications
About this item
Highly effective skin care cream (W/O emulsion) for extremely stressed and very dry skin. Can be applied to hands and face.
- Lipid level of approximately 30 %
- Without parabens, silicone-free and perfumed
- Helps to support the regeneration process of damaged skin, if applied regularly
- Contains moisture-binding substance sorbitol as well as isopropyl myristate that functions as a lipid-replenishing agent
- Helps to foster the regeneration process when the skin suffers from dermatitis or eczema
- Recommended as concomitant skin care for the therapy of irritated skin
The cream increases the skin's ability to retain water, supports the regeneration of the skin's natural protective layer and improves the skin's smoothness.