DX4™ Zip Lightweight Mid Layer Tops
Informazioni su questo articolo
The DX4 zip base layer top is lightweight and breathable, which assists with perfect body temperature control. The high stretch, highly durable nylon fabric complements the body mapped design. The full front zip with chin guard has a rubber puller for easy grip.
- 4-way stretch fabric for ease of movement and added comfort
- Engineered with an active fit
- Raglan sleeves for fluid movement and comfort
- Internal pockets for safe storage
- Zipped chest pocket
- 6 pockets for ample storage
- Side zip pockets
- Contemporary design with an ergonomic cut
- Contrast colouring for added style
- Easy grip pullers attached to all zips
- Bartacked at all stress points
The DX4 zip lightweight mid layer top features zipped hand and chest pockets that are ideal for securely storing smaller items, elasticated cuffs for a secure fit, thumb loops, a curved back hem for added protection, contrast premium zips for added style, and a front zip opening for easy access.
Colour: Black or metro blue
Weather conditions - Dry: Daily wear
7 varianti
Product Details & Documents
The DX4 zip base layer top is lightweight and breathable, which assists with perfect body temperature control. The high stretch, highly durable nylon fabric complements the body mapped design. The full front zip with chin guard has a rubber puller for easy grip.
- 4-way stretch fabric for ease of movement and added comfort
- Engineered with an active fit
- Raglan sleeves for fluid movement and comfort
- Internal pockets for safe storage
- Zipped chest pocket
- 6 pockets for ample storage
- Side zip pockets
- Contemporary design with an ergonomic cut
- Contrast colouring for added style
- Easy grip pullers attached to all zips
- Bartacked at all stress points
The DX4 zip lightweight mid layer top features zipped hand and chest pockets that are ideal for securely storing smaller items, elasticated cuffs for a secure fit, thumb loops, a curved back hem for added protection, contrast premium zips for added style, and a front zip opening for easy access.
Colour: Black or metro blue
Weather conditions - Dry: Daily wear
Specifications for Product Family
Specifiche
- Catalog No:
- PRTWDX480BKRS
- PRTWDX480BKRM
- PRTWDX480BKRL
- PRTWDX480BKRXL
- PRTWDX480BKRXXL
- PRTWDX480BKRXXXL
- PRTWDX480MBRL
- Applicazioni per abiti:
- Casual|Outdoor
- Casual|Outdoor
- Casual|Outdoor
- Casual|Outdoor
- Casual|Outdoor
- Casual|Outdoor
- Casual|Outdoor
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Genere:
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoVestibilità del capoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRSApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRMApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRLApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRXLApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRXXLApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480BKRXXXLApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex - Specifiche:
More
Specifications
Cat. No.PRTWDX480MBRLApplicazioni per abitiCasual|OutdoorRisk areaColdGenerePer l'uomoSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoNylon/Spandex