BioCleanPlate Wako™ Microplates
Catalog # WAKF293-35221
Anbieter: FUJIFILM WAKO CHEMICALS
Technische Daten
- Descrizione:BioCleanPlate Wako™, 50×96-wells/plate, <0,01 EU/plate
- Conf:50
Technische Daten
Über diesen Artikel
The BioCleanPlate Wako™ are endotoxin-free microplates that are individually wrapped to reduce the chance of contamination and are perfect for LAL testing.
- Endotoxin-free 96-well microplate (<0,01 EU/plate)
- Individually packaged, ideal for use in clean rooms
- Packaging film features dust-free protection
- Plates are designed for use with microplate readers
When used in conjunction with BioCleanTip Wako® Extend S allows for aliquoting multi-test reagent into Microplates accurately and precisely avoiding error or bubbles.