Orders LinkContactUsLinkComponent
Unbreakable Storage Boxes
Unbreakable Storage Boxes
Supplier:   MULTIROIR
Unbreakable Storage Boxes
Supplier:  MULTIROIR
Certificates
Unbreakable Storage Boxes
Unbreakable Storage Boxes
3 Options Available Below
Product Details & Documents
This series of storage boxes appeals to both private and professional users for its versatility, space-saving when empty, simplicity, and robust design.
  • Excellent load capacity
  • Stackable and built-in
  • Shock and impact-resistant
  • Excellent lid retention by the closing clips
  • BPA and Phthalate-free
These unbreakable storage boxes are specially designed to meet the needs of professionals. They offer versatile use and space optimisation. This series comes in 3 distinct models, each equipped with excellent load capacity. The bins are designed to stack on top of each other and can also interlock to save space. Their robust construction makes them resistant to shocks and impacts, ensuring optimal protection for the contents. The closure clips ensure an excellent lid grip, preventing any risk of accidental opening.
These bins can withstand temperatures ranging from ‒40 to 100 °C, making them suitable for a wide range of environments.
Product Family Options
Product Information
  • Capacità
    Colore
    L×P×A
    Materiale
    Availability
    Price
  • 14 L
    nero
    External: 360×270 mm
    Internal: 325×245 mm
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 311.00
    SFr. 311.00
    1 * 10 ST
       SFr. 311.00
  • 32 L
    nero
    External: 450×340 mm
    Internal: 395×300 mm
    PP
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - SFr. 371.00
    SFr. 371.00
    1 * 6 ST
       SFr. 371.00
  • 50 L
    nero
    External: 530×344 mm
    Internal: 475×290 mm
    PP
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - SFr. 318.00
    SFr. 318.00
    1 * 4 ST
       SFr. 318.00