Orders LinkContactUsLinkComponent
Storage Boxes, 100% Recycled Polypropylene
 :   MULTIROIR
Storage Boxes, 100% Recycled Polypropylene
 :  MULTIROIR
Storage Boxes, 100% Recycled Polypropylene
Storage Boxes, 100% Recycled Polypropylene
This range consists of four storage bins, all made from 100% recycled polypropylene.
  • Solid, stackable, and palletisable storage bin made of polypropylene
  • Compliant with european standard for easy storage and transportation
  • Easy handling with open handles
  • Manufactured from recycled polypropylene
  • Lightweight and durable
  • Heights from 120 to 320 mm
These multifunctional storage bins are highly durable and suitable for all types of use. The entire range is certified BPA-free.
Product Family Options
  • Capacità
    Colore
    L×P×A
    Materiale
  • 12,5 L
    nero
    External: 390×290×170 mm
    Internal: 350×250 mm
    PP
    1 * 12 ST
    1 * 12 ST - SFr. 186.00
    SFr. 186.00
    1 * 12 ST
       SFr. 186.00
  • 32 L
    nero
    External: 390×390×265 mm
    Internal: 350×350 mm
    PP
    1 * 12 ST
    1 * 12 ST - SFr. 273.00
    SFr. 273.00
    1 * 12 ST
       SFr. 273.00
  • 60 L
    nero
    External: 580×390×350 mm
    Internal: 540×360 mm
    PP
    1 * 6 ST
    1 * 6 ST - SFr. 350.00
    SFr. 350.00
    1 * 6 ST
       SFr. 350.00
  • 75 L
    nero
    External: 760×390×350 mm
    Internal: 720×350 mm
    PP
    1 * 6 ST
    1 * 6 ST - SFr. 230.00
    SFr. 230.00
    1 * 6 ST
       SFr. 230.00