Orders LinkContactUsLinkComponent
Sample Vials, with Snap-Cap
Sample Vials, with Snap-Cap
Sample Vials, with Snap-Cap
Sample Vials, with Snap-Cap
Soda-lime glass, amber.
  • Amber glass especially for light sensitive liquids
  • White snap-caps are impervious to ultraviolet light
  • Snap-caps included
Ideal for storing medication as liquid, powder or capsules.
Product Family Options
  • Capacità
    Ø×A
    Materiale
    Colore
    Tappo incluso
    Tipologia tappo
    Dimensione tappo
    Materiale tappo
    Colore tappo
    Sterili
  • 5,00 ml
    19×40 mm
    Vetro
    ambra
    Yes
    Snap cap, closed top
    20 mm
    PE
    bianco
    -
    1 * 200 ST
    1 * 200 ST - SFr. 86.80
    SFr. 86.80
    1 * 200 ST
       SFr. 86.80
  • 10,00 ml
    22×45 mm
    Vetro
    ambra
    Yes
    Snap cap, closed top
    22 mm
    PE
    bianco
    -
    1 * 200 ST
    1 * 200 ST - SFr. 98.10
    SFr. 98.10
    1 * 200 ST
       SFr. 98.10
  • 25,00 ml
    30×50 mm
    Vetro
    ambra
    Yes
    Snap cap, closed top
    30 mm
    PE
    bianco
    -
    1 * 200 ST
    1 * 200 ST - SFr. 144.00
    SFr. 144.00
    1 * 200 ST
       SFr. 144.00
  • 30,00 ml
    28×75 mm
    Vetro
    ambra
    Yes
    Snap cap, closed top
    28 mm
    PE
    bianco
    -
    1 * 200 ST
    1 * 200 ST - SFr. 141.00
    SFr. 141.00
    1 * 200 ST
       SFr. 141.00