Plastic Box, Translucent Polypropylene
Informazioni su questo articolo
This range of plastic boxes offers a modern and practical solution for the storage, organization, transportation, and presentation of small parts. With their innovative design, these plastic boxes are ideal for various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
- 5 available formats: A4, A5, A6, A7, and A8
- Material: 100% recyclable virgin polypropylene
- Hinged lid with secure snap closure
- Excellent stackability due to stacking feet
- Temperature resistance from –20 to +80 °C
- Clear lid for excellent content visibility
- Dividers reaching the top of the box to prevent movement of small items between compartments
- Fixed and removable compartments for A4, A5, and A6 formats, fixed compartments for A7 and A8 formats
Their versatility makes them ideal for many applications in various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
12 varianti
Product Details & Documents
This range of plastic boxes offers a modern and practical solution for the storage, organization, transportation, and presentation of small parts. With their innovative design, these plastic boxes are ideal for various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
- 5 available formats: A4, A5, A6, A7, and A8
- Material: 100% recyclable virgin polypropylene
- Hinged lid with secure snap closure
- Excellent stackability due to stacking feet
- Temperature resistance from –20 to +80 °C
- Clear lid for excellent content visibility
- Dividers reaching the top of the box to prevent movement of small items between compartments
- Fixed and removable compartments for A4, A5, and A6 formats, fixed compartments for A7 and A8 formats
Their versatility makes them ideal for many applications in various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
Product Family Options
Informazioni sul prodotto
- L×P×ADisponibilitàPrice
- 216-3024Plastic box, 1 compartment, compartment size 82×53 mmPlastic box, 1 compartment, compartment size 82×53 mm90×60×20 mmLog in per disponibilità1 * 24 STSFr. 45.50
- 216-3025Plastic box, 2 compartments, compartment size 53×40 mmPlastic box, 2 compartments, compartment size 53×40 mm90×60×20 mmLog in per disponibilità1 * 24 STSFr. 51.40
- 216-3026Plastic box, 4 compartments, compartment size 40×26 mmPlastic box, 4 compartments, compartment size 40×26 mm90×60×20 mmLog in per disponibilità1 * 24 STSFr. 51.40
- 216-3027Plastic box, 6 compartments, compartment size 26×26 mmPlastic box, 6 compartments, compartment size 26×26 mm90×60×20 mmLog in per disponibilità1 * 24 STSFr. 51.40
- 216-3028Plastic box, 1 compartment, compartment size 121×72 mmPlastic box, 1 compartment, compartment size 121×72 mm130×90×25 mmLog in per disponibilità1 * 16 STSFr. 41.20
- 216-3029Plastic box, 12 compartments, compartment size 29×26 mmPlastic box, 12 compartments, compartment size 29×26 mm130×90×25 mmLog in per disponibilità1 * 16 STSFr. 44.00
- 216-3030Plastic box, 1 compartment, compartment size 171×118 mmPlastic box, 1 compartment, compartment size 171×118 mm190×130×30 mmLog in per disponibilità1 * 16 STSFr. 59.10
- 216-3031Plastic box, 12 compartments, compartment size 40×38 mmPlastic box, 12 compartments, compartment size 40×38 mm190×130×30 mmLog in per disponibilità1 * 16 STSFr. 71.00
- 216-3032Plastic box, 1 compartment, compartment size 250×153 mmPlastic box, 1 compartment, compartment size 250×153 mm260×180×40 mmLog in per disponibilità1 * 14 STSFr. 99.80
- 216-3033Plastic box, 24 compartments, compartment size 39×38 mmPlastic box, 24 compartments, compartment size 39×38 mm260×180×40 mmLog in per disponibilità1 * 14 STSFr. 115.00
- 216-3034Plastic box, 1 compartment, compartment size 351×239 mmPlastic box, 1 compartment, compartment size 351×239 mm360×260×50 mmLog in per disponibilità1 * 6 STSFr. 84.70
- 216-3035Plastic box, 28 compartments, compartment size 59×45 mmPlastic box, 28 compartments, compartment size 59×45 mm360×260×50 mmLog in per disponibilità1 * 6 STSFr. 96.90