Janitorial Cart
Codice catalogo 129-2176
Nome del fornitore: Rubbermaid Commercial Products
Specifiche
- Colore:nero
- Descrizione:Janitorial cart
- L×P×A:552×1168×975 mm
- Conf:1
Specifiche
Informazioni su questo articolo
The traditional janitorial cleaning cart is lightweight and maneuverable to collect waste and transports tools for efficient cleaning.
- Water and chemical resistant surface provide easy maintenance and long-lasting good looks
- Front-access zip bag allows user to easily remove collected waste with no need to lift through the top
- Front platform holds up to a 121 L BRUTE® container
- Two fixed casters/two swivel wheels
Eleven tool hooks make room for all your cleaning needs, such as brooms, mops, dust pans, dusters, and safety signs.
Delivery: Supplied with yellow vinyl bag (92 L).