Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Quick-RNA™ Viral Kit

Quick-RNA™ Viral Kit

Quick-RNA™ viral kit, 200 Preps, 1 KIT
Codice catalogo ZYMOR1035
Marketsource
Nome del fornitore:   Zymo Research
Quick-RNA™ Viral Kit
Codice catalogo ZYMOR1035
Nome del fornitore:  Zymo Research
Marketsource
Numero CAS:  
Quick-RNA™ Viral Kit
Quick-RNA™ Viral Kit

Quick-RNA™ viral kit, 200 Preps, 1 KIT

Numero CAS:  
Price
SFr. 703.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents

The Quick-RNA™ viral kit is a quick, purification system for viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield™.

  • Quick, spin-column purification of viral RNA from plasma, serum, CSF, cell culture media, cellular suspensions, urine, blood, saliva, swab, fecal
  • RNA is ready for Next-Gen sequencing, RT/PCR, hybridisation
  • DNA/RNA Shield™ is included for nucleic acid stability during sample storage/transport at ambient temperatures

DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures (4 - 25 °C). The reagent effectively lyses cells and inactivates nucleases and infectious agents (virus).

The kit also features a specialized buffer system that facilitates complete viral particle lysis for efficient RNA isolation from samples containing enteroviruses, rhinoviruses, coronaviruses, HIV, HCV, influenza A virus, flaviviruses, measles virus, parainfluenza virus, parvovirus (a ssDNA virus), etc. Viral RNA is bound to the column, washed and eluted. The isolated high-quality viral RNA is ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/PCR detection.

Informazioni per l'ordine: The kit includes DNA/RNA Shield™ (2X concentrate), viral RNA buffer, viral wash buffer (concentrate), DNase/RNase-free water, Zymo-Spin™ IC columns, collection tubes and instruction manual.

Specifiche

Specifiche

  • Conf:
    1 KIT
  • Descrizione:
    Quick-RNA™ viral kit
  • No. of reactions:
    200 Preps
  • Cat. No.:
    ZYMOR1035
  • Prep size:
    Mini
  • Refine target molecule:
    Viral RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Blood|Biofluids (serum, plasma, saliva)
  • Descrizione:
    Quick-RNA™ Viral Kit

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..