Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Quick-RNA™ Plant Kit

Quick-RNA™ plant kit, 50 Preps, 1 KIT
  ZYMOR2024
Marketsource
Quick-RNA™ Plant Kit
  ZYMOR2024
Marketsource
 :  
Quick-RNA™ Plant Kit
Quick-RNA™ Plant Kit

Quick-RNA™ plant kit, 50 Preps, 1 KIT

 :  
 
SFr. 424.00
   
1 * 1 KIT
 

Quick-RNA plant miniprep kit is designed for rapid isolation of RNA from various plant samples such as leaves, stems, buds, flowers, fruits and seeds.

  • Extract intact high-quality total RNA (including small/micro RNAs) from a wide variety of plant samples
  • Ultra-high density bashing bead technology for complete disruption and sample lysis
  • Inhibitor free RNA is ready for all downstream applications
  • Size Range Total RNA ≥ 17 nt

The Zymo-Spin IIICG column allows for high-capacity DNA elimination and the subsequent Zymo-Spin IIC column efficiently adsorbs total RNA. The RNA is washed and then eluted with DNase/RNase free water. For inhibitor removal, the eluted RNA can be treated by running the sample through the Zymo-Spin III-HRC spin filters. The RNA is suitable for use in various subsequent procedures including sequencing, RT-PCR and microarray.

 

 

  • Conf:
    1 KIT
  • Descrizione:
    Quick-RNA™ plant kit
  • No. of reactions:
    50 Preps
  • Cat. No.:
    ZYMOR2024
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Plant samples
  • Resa:
    50 µg RNA (binding capacity), ≥25 µl (elution volume)
  • Descrizione:
    Quick-RNA plant kit

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.