EC (Escherichia coli) Broth acc. ISO 7251, FDA‑BAM and SMWW GranuCult® plus, Millipore
Catalog # 1.10765.0500
Supplier: Merck
CAS Number:
Specifications
- Tipo di conf.:Plastic bottle
- Conf:500 g
- Descrizione:EC Broth GranuCult® plus
Specifications
About this item
EC (Escherichia coli) broth
- Granulation preserves homogeneity
- Easy to weigh out
- Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
Media is granulated.