High-speed barcode reader, VisionMate™ HSX
Product Details & Documents
The Thermo Scientific™ VisionMate™ HSX High Speed Barcode Reader decodes 2D tube barcodes in as little as one second. Compatible with Matrix™ and Nunc™ tubes in 24, 48, 96, and 384 format racks.
- Pre-loaded rack configurations allow you to rapidly read Matrix™ and Nunc™ racks of tubes without additional set up steps
- Can decode both 2D and linear rack barcodes, providing additional tracking options
- Customizable LED visual feedback provides the user with instant notification on successful reads, as well as notification of any read errors
- Included magnetic linear barcode reader can be positioned in various locations on the all metal housing of the base unit to read linear rack barcodes.
- Cyroprotection provides resistance to the accumulation of frost on the reading surface
- Base unit is fully powered by a single USB connection to your computer
- Can be used on the benchtop or integrated into your automated system
Supported Barcode Types: All Thermo Fisher Matrix™ and Nunc™ 2D Barcoded Tubes, ECC200, ISO 16022:2006, Linear Barcodes
The redesigned VisionMate™ Suite software provides a range of export options for seamless transfers of barcode data to your LIMS, ELN, or database.
The VisionMate™ HSX carries an IP66 rating, which protects the instrument from various adverse environmental conditions in your laboratory and establishes the critical environment barrier needed for use in a cleanroom.
A perfect complement to the Thermo Scientific™ Matrix™ and Nunc™ 2D barcoded tube offerings, the VisionMate™ HSX provides valuable process flexibility and compatibility in storage workflows in drug discovery, biological, or genomic storage environments.
Specifiche
- Descrizione:High-speed barcode reader, VisionMate™ HSX
- Capacità memoria:No internal storage capacity; data is stored on the user's computer
- Cat. No.:479-2448
- Interfaccia:USB 2.0+
- Potenza:USB Powered
- Environmentally Preferable:Produzione a basso impatto
- L×P×A:200×150×160 mm
- Certificazione ambientale:NAR Certificate of Retirement