Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Catalog # 114-9596
Supplier: ISOFIELD IRELAND
Specifications
- Tipo:With ties
- Colore:bianco
- Taglia:Universal
- Conf:500
Specifications
About this item
The Isofield Zetta facemask is ISO Class 4 compatible.
- ISO Class 4 compatible
- Extra wide at 230 mm
- Four ties made from polyester
- Individually packed
- Low CFU
- Excellent BFE and PFE
- Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable polyester ties, the Zetta is made from low-linting cleanroom grade material to ensure best in class cleanliness. The individually packed Zetta facemasks provide excellent bacterial and particle filtration efficiency.
Packaging: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 10 inner bags per case (500 pieces).