Elma Sinks
Product Details & Documents
Elma Sink is simply reliable, providing a step ahead to ensure items are thoroughly rinsed.
- For immersion rinsing or rinsing off objects
- Stainless steel tank
- Elma Sink L and XL sizes are equipped with a drain
- Bath sizes correspond to Elmasonic Easy, Med, Select and P units
For perfect cleaning results. The Elma Sink is an important addition to the overall cleaning process. This tabletop unit is available in four different sizes – the L and XL models feature an additional side drain. Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink. This ensures that any residues and cleaning chemicals are rinsed off – thoroughly. In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results.
Informazioni per l'ordine: All units are supplied with operating instructions, Sink X and XL units are supplied with a hose end and hose clamp.
Specifiche
- Descrizione:Elma Sink S
- Seq:01
- Cat. No.:ELMA1113839
- Peso:2,0 kg
- Modello:Elma Sink S
- Dimensioni della vasca L×P×A:215×115×145 mm
- Capacità:3,9 L
- L×P×A:305×155×220 mm