VascuLife® VEGF-MV Microvascular Medium, Lifeline Cell Technology
Catalog # CELSLL-0005
Supplier: CELL SYSTEMS
CAS Number:
Specifications
- Conf:1 KIT
- Descrizione:VascuLife® VEGF-Mv Medium Complete Kit
Specifications
About this item
VascuLife® VEGF-Mv is optimized for the culture of human microvascular endothelial cells, and supports the rapid proliferation of these cells in a 5% serum environment.
- Optimised for the culture of human microvascular endothelial cells
- Supports rapid cell proliferation
- Contains no antimicrobials or phenol red
The VascuLife® VEGF-Mv LifeFactors® kit includes: rh FGF basic 0,5 ml, Ascorbic acid 0,5 ml, Hydrocortisone hemisuccinate 0,5 ml, L-Glutamine 25 ml, rh IGF-1 0,5 ml, rh EGF 0,5 ml, rh VEGF 0,5 ml, Heparin sulfare 0,5 ml, FBS 25 ml, Gentamicin amphotericin B 0,5 ml.
Ordering information: The VascuLife® VEGF-Mv LifeFactors® kit includes: rh FGF basic 0,5 ml, Ascorbic acid 0,5 ml, Hydrocortisone hemisuccinate 0,5 ml, L-Glutamine 25 ml, rh IGF-1 0,5 ml, rh EGF 0,5 ml, rh VEGF 0,5 ml, Heparin sulfare 0,5 ml, FBS 25 ml, Gentamicin amphotericin B 0,5 ml.