Cole-Parmer® ST-200 and SHP-200 Series 3-Position Magnetic Stirrers
Three independent stirring or heating/stirring positions yet only measures L×W: 600×270 mm.
- Powerful magnets for strong magnetic coupling and stirring capacity
- Easily accommodate a 2 L beaker in each position
Three independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.
: Supplied with three 25 mm PTFE coated magnetic stirring bars and power cords with UK and Shuko plug.
- Catalog No:
- STUA51400-02
- STUA51600-08
- Modello:
- -
- SHP-200-MP
- Numero di posizioni di agitazione:
- 3
- 3
- Intervallo di velocità:
- 0 - 1500 min⁻¹
- 100 - 1500 min⁻¹
- Dimensioni della piastra:
- 120×550 mm
- 160×160 mm
- L×P×A:
- 270×600×120 mm
- 270×600×110 mm
- Elettrico:
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
Frequently Bought Together